RIE156320925 Rieke Metals, Inc.

4'-Fluoro-4-iodo-3'-methylbenzophenone

  • MDL number MFCD09801648
  • Molecular Weight : 340.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5209-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5209-22
Availability details for
Quantity
  • Availability
  • Add to Cart