RIE156320929 Rieke Metals, Inc.

2-Chloro-4'-isopropylbenzophenone

  • MDL number MFCD09801654
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl)(4-isopropylphenyl)methanone
Inchi Code 1S/C16H15ClO/c1-11(2)12-7-9-13(10-8-12)16(18)14-5-3-4-6-15(14)17/h3-11H,1-2H3
Inchi Key MFXGAJQJEQICNK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5210-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5210-14
Availability details for
Quantity
  • Availability
  • Add to Cart