RIE156320931 Rieke Metals, Inc.

2-Iodo-4'-isopropylbenzophenone

  • MDL number MFCD07775692
  • Molecular Weight : 350.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5210-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5210-20
Availability details for
Quantity
  • Availability
  • Add to Cart