RIE156320935 Rieke Metals, Inc.

3-Chloro-2'-methylbenzophenone

  • MDL number MFCD09801658
  • Cas Number : 5621-69-2
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5211-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5211-15
Availability details for
Quantity
  • Availability
  • Add to Cart