RIE156320941 Rieke Metals, Inc.

4-Iodo-2'-methylbenzophenone

  • MDL number MFCD09801664
  • Molecular Weight : 322.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-iodophenyl)(2-methylphenyl)methanone
Inchi Code 1S/C14H11IO/c1-10-4-2-3-5-13(10)14(16)11-6-8-12(15)9-7-11/h2-9H,1H3
Inchi Key FSXKCMMRABVKHA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5211-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5211-22
Availability details for
Quantity
  • Availability
  • Add to Cart