RIE156320942 Rieke Metals, Inc.

4-tert-Butyl-2'-methylbenzophenone

  • MDL number MFCD09801665
  • Cas Number : 14252-16-5
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-tert-butylphenyl)(2-methylphenyl)methanone
Inchi Code 1S/C18H20O/c1-13-7-5-6-8-16(13)17(19)14-9-11-15(12-10-14)18(2,3)4/h5-12H,1-4H3
Inchi Key GRJQUSDNXCHZTG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5211-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5211-34
Availability details for
Quantity
  • Availability
  • Add to Cart