RIE156320953 Rieke Metals, Inc.

2-Iodo-4'-methylbenzophenone

  • MDL number MFCD00767183
  • Cas Number : 107624-39-5
  • Molecular Weight : 322.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5213-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5213-20
Availability details for
Quantity
  • Availability
  • Add to Cart