RIE156320959 Rieke Metals, Inc.

2-Bromo-4'-n-pentylbenzophenone

  • MDL number MFCD09801679
  • Cas Number : 64358-18-5
  • Molecular Weight : 331.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5214-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5214-17
Availability details for
Quantity
  • Availability
  • Add to Cart