RIE156320961 Rieke Metals, Inc.

4-Bromo-4'-n-pentylbenzophenone

  • MDL number MFCD09801681
  • Cas Number : 918500-19-3
  • Molecular Weight : 331.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromophenyl)(4-pentylphenyl)methanone
Inchi Code 1S/C18H19BrO/c1-2-3-4-5-14-6-8-15(9-7-14)18(20)16-10-12-17(19)13-11-16/h6-13H,2-5H2,1H3
Inchi Key VKNWSBODGBUHBL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5214-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5214-19
Availability details for
Quantity
  • Availability
  • Add to Cart