RIE156320962 Rieke Metals, Inc.

2-Iodo-4'-n-pentylbenzophenone

  • MDL number MFCD09801682
  • Cas Number : 64358-28-7
  • Molecular Weight : 378.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-iodophenyl)(4-pentylphenyl)methanone
Inchi Code 1S/C18H19IO/c1-2-3-4-7-14-10-12-15(13-11-14)18(20)16-8-5-6-9-17(16)19/h5-6,8-13H,2-4,7H2,1H3
Inchi Key OJWHCVHKKDVTIM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5214-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5214-20
Availability details for
Quantity
  • Availability
  • Add to Cart