RIE156320963 Rieke Metals, Inc.

3-Iodo-4'-n-pentylbenzophenone

  • MDL number MFCD09801683
  • Molecular Weight : 378.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5214-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5214-21
Availability details for
Quantity
  • Availability
  • Add to Cart