RIE156320964 Rieke Metals, Inc.

4-Iodo-4'-n-pentylbenzophenone

  • MDL number MFCD09801684
  • Molecular Weight : 378.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-iodophenyl)(4-pentylphenyl)methanone
Inchi Code 1S/C18H19IO/c1-2-3-4-5-14-6-8-15(9-7-14)18(20)16-10-12-17(19)13-11-16/h6-13H,2-5H2,1H3
Inchi Key GCQDCVBJLDQYRV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5214-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5214-22
Availability details for
Quantity
  • Availability
  • Add to Cart