RIE156320967 Rieke Metals, Inc.

3,5-Difluoro-4'-n-pentylbenzophenone

  • MDL number MFCD09801687
  • Molecular Weight : 288.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5214-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5214-42
Availability details for
Quantity
  • Availability
  • Add to Cart