RIE156320970 Rieke Metals, Inc.

2-Chloro-4'-n-propylbenzophenone

  • MDL number MFCD09801690
  • Cas Number : 64358-07-2
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5215-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5215-14
Availability details for
Quantity
  • Availability
  • Add to Cart