RIE156320971 Rieke Metals, Inc.

2-Bromo-4'-n-propylbenzophenone

  • MDL number MFCD09801691
  • Cas Number : 64358-16-3
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromophenyl)(4-propylphenyl)methanone
Inchi Code 1S/C16H15BrO/c1-2-5-12-8-10-13(11-9-12)16(18)14-6-3-4-7-15(14)17/h3-4,6-11H,2,5H2,1H3
Inchi Key XZJWLLALPZMUJJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5215-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5215-17
Availability details for
Quantity
  • Availability
  • Add to Cart