RIE156320975 Rieke Metals, Inc.

4-tert-Butyl-4'-n-propylbenzophenone

  • MDL number MFCD09801692
  • Molecular Weight : 280.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5215-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5215-34
Availability details for
Quantity
  • Availability
  • Add to Cart