RIE156320979 Rieke Metals, Inc.

4-Chloro-2'-(thiomethyl)benzophenone

  • MDL number MFCD09801696
  • Molecular Weight : 262.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5216-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5216-16
Availability details for
Quantity
  • Availability
  • Add to Cart