RIE156320983 Rieke Metals, Inc.

4-tert-Butyl-2'-(thiomethyl)benzophenone

  • MDL number MFCD09801700
  • Molecular Weight : 284.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5216-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5216-34
Availability details for
Quantity
  • Availability
  • Add to Cart