RIE156320984 Rieke Metals, Inc.

3,4-Difluoro-2'-(thiomethyl)benzophenone

  • MDL number MFCD09801701
  • Molecular Weight : 264.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-difluorophenyl)[2-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C14H10F2OS/c1-18-13-5-3-2-4-10(13)14(17)9-6-7-11(15)12(16)8-9/h2-8H,1H3
Inchi Key GNFUAPRPKBHEDJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5216-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5216-41
Availability details for
Quantity
  • Availability
  • Add to Cart