RIE156320986 Rieke Metals, Inc.

3,4-Dichloro-2'-(thiomethyl)benzophenone

  • MDL number MFCD09801703
  • Molecular Weight : 297.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5216-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5216-47
Availability details for
Quantity
  • Availability
  • Add to Cart