RIE156320987 Rieke Metals, Inc.

3,5-Dichloro-2'-(thiomethyl)benzophenone

  • MDL number MFCD09801704
  • Molecular Weight : 297.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dichlorophenyl)[2-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C14H10Cl2OS/c1-18-13-5-3-2-4-12(13)14(17)9-6-10(15)8-11(16)7-9/h2-8H,1H3
Inchi Key TXYWZXASMHXKIF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5216-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5216-48
Availability details for
Quantity
  • Availability
  • Add to Cart