RIE156320988 Rieke Metals, Inc.

2-Chloro-4'-(thiomethyl)benzophenone

  • MDL number MFCD09801705
  • Cas Number : 55270-72-9
  • Molecular Weight : 262.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5217-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5217-14
Availability details for
Quantity
  • Availability
  • Add to Cart