RIE156320990 Rieke Metals, Inc.

4-tert-Butyl-4'-(thiomethyl)benzophenone

  • MDL number MFCD09801707
  • Cas Number : 73242-05-4
  • Molecular Weight : 284.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-tert-butylphenyl)[4-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C18H20OS/c1-18(2,3)15-9-5-13(6-10-15)17(19)14-7-11-16(20-4)12-8-14/h5-12H,1-4H3
Inchi Key ZSHWCYKHLQGNJV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5217-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5217-34
Availability details for
Quantity
  • Availability
  • Add to Cart