RIE156320991 Rieke Metals, Inc.

3,5-Dichloro-4'-(thiomethyl)benzophenone

  • MDL number MFCD09801708
  • Cas Number : 197439-14-8
  • Molecular Weight : 297.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5217-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5217-48
Availability details for
Quantity
  • Availability
  • Add to Cart