RIE156320994 Rieke Metals, Inc.

4-Bromo-3',4',5'-trifluorobenzophenone

  • MDL number MFCD09801711
  • Molecular Weight : 315.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5218-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5218-19
Availability details for
Quantity
  • Availability
  • Add to Cart