RIE156320997 Rieke Metals, Inc.

4-Iodo-3',4',5'-trifluorobenzophenone

  • MDL number MFCD09801714
  • Molecular Weight : 362.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5218-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5218-22
Availability details for
Quantity
  • Availability
  • Add to Cart