RIE156320998 Rieke Metals, Inc.

4-tert-Butyl-3',4',5'-trifluorobenzophenone

  • MDL number MFCD09801715
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-tert-butylphenyl)(3,4,5-trifluorophenyl)methanone
Inchi Code 1S/C17H15F3O/c1-17(2,3)12-6-4-10(5-7-12)16(21)11-8-13(18)15(20)14(19)9-11/h4-9H,1-3H3
Inchi Key FMHSSWRKFUEMHN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5218-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5218-34
Availability details for
Quantity
  • Availability
  • Add to Cart