RIE156320999 Rieke Metals, Inc.

3,4-Difluoro-3',4',5'-trifluorobenzophenone

  • MDL number MFCD09801716
  • Molecular Weight : 272.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-difluorophenyl)(3,4,5-trifluorophenyl)methanone
Inchi Code 1S/C13H5F5O/c14-8-2-1-6(3-9(8)15)13(19)7-4-10(16)12(18)11(17)5-7/h1-5H
Inchi Key IEBDIDHXDAZTNS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5218-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5218-41
Availability details for
Quantity
  • Availability
  • Add to Cart