RIE156321000 Rieke Metals, Inc.

3,5-Difluoro-3',4',5'-trifluorobenzophenone

  • MDL number MFCD09801717
  • Molecular Weight : 272.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5218-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5218-42
Availability details for
Quantity
  • Availability
  • Add to Cart