RIE156321003 Rieke Metals, Inc.

4'-(1,3-Dioxan-2-yl)-2-trifluoromethylbenzophenone

  • MDL number MFCD03844157
  • Molecular Weight : 336.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5219-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5219-08
Availability details for
Quantity
  • Availability
  • Add to Cart