RIE156321004 Rieke Metals, Inc.

2-Chloro-3-(2-ethoxycarbonylbenzoyl)pyridine

  • MDL number MFCD07699002
  • Cas Number : 890100-43-3
  • Molecular Weight : 289.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-[(2-chloro-3-pyridinyl)carbonyl]benzoate
Inchi Code 1S/C15H12ClNO3/c1-2-20-15(19)11-7-4-3-6-10(11)13(18)12-8-5-9-17-14(12)16/h3-9H,2H2,1H3
Inchi Key AUZCWXKJWVICAP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5500

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5500
Availability details for
Quantity
  • Availability
  • Add to Cart