RIE156321006 Rieke Metals, Inc.

2-chloro-3-(4-ethoxycarbonylbenzoyl)pyridine

  • MDL number MFCD02260447
  • Cas Number : 760192-98-1
  • Molecular Weight : 289.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5502

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5502
Availability details for
Quantity
  • Availability
  • Add to Cart