RIE156321008 Rieke Metals, Inc.

2-chloro-3-(4-cyanobenzoyl)pyridine

  • MDL number MFCD07699004
  • Cas Number : 890100-45-5
  • Molecular Weight : 242.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(2-chloro-3-pyridinyl)carbonyl]benzonitrile
Inchi Code 1S/C13H7ClN2O/c14-13-11(2-1-7-16-13)12(17)10-5-3-9(8-15)4-6-10/h1-7H
Inchi Key KJXQJPSYRHMEPU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5507

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5507
Availability details for
Quantity
  • Availability
  • Add to Cart