RIE156321010 Rieke Metals, Inc.

2-Chloro-5-(2-ethoxycarbonylbenzoyl)pyridine

  • MDL number MFCD02260457
  • Cas Number : 890100-47-7
  • Molecular Weight : 289.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-[(6-chloro-3-pyridinyl)carbonyl]benzoate
Inchi Code 1S/C15H12ClNO3/c1-2-20-15(19)12-6-4-3-5-11(12)14(18)10-7-8-13(16)17-9-10/h3-9H,2H2,1H3
Inchi Key HGDSAUTYEMJTPJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5512

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5512
Availability details for
Quantity
  • Availability
  • Add to Cart