RIE156321011 Rieke Metals, Inc.

2-chloro-5-(3-ethoxycarbonylbenzoyl)pyridine

  • MDL number MFCD02260458
  • Cas Number : 890100-48-8
  • Molecular Weight : 289.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5513

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5513
Availability details for
Quantity
  • Availability
  • Add to Cart