RIE156321013 Rieke Metals, Inc.

2-Chloro-3-(5-ethoxycarbonyl-2-thenoyl)pyridine

  • MDL number MFCD02260470
  • Cas Number : 890100-49-9
  • Molecular Weight : 295.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-[(2-chloro-3-pyridinyl)carbonyl]-2-thiophenecarboxylate
Inchi Code 1S/C13H10ClNO3S/c1-2-18-13(17)10-6-5-9(19-10)11(16)8-4-3-7-15-12(8)14/h3-7H,2H2,1H3
Inchi Key IBKZUXHYWFOQNZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5525

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5525
Availability details for
Quantity
  • Availability
  • Add to Cart