RIE156321016 Rieke Metals, Inc.

ethyl 5-(2-thenoyl)-2-furoate

  • MDL number MFCD02260477
  • Cas Number : 890100-52-4
  • Molecular Weight : 250.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-(2-thienylcarbonyl)-2-furoate
Inchi Code 1S/C12H10O4S/c1-2-15-12(14)9-6-5-8(16-9)11(13)10-4-3-7-17-10/h3-7H,2H2,1H3
Inchi Key HCHCMBBYLXNJGL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5532

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5532
Availability details for
Quantity
  • Availability
  • Add to Cart