RIE156321018 Rieke Metals, Inc.

Ethyl 5-(2-furanoyl)-2-furoate

  • MDL number MFCD02260479
  • Cas Number : 890100-54-6
  • Molecular Weight : 234.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5534

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5534
Availability details for
Quantity
  • Availability
  • Add to Cart