RIE156321023 Rieke Metals, Inc.

Ethyl 4-(6-chloropyridin-3-yl)-4-oxobutyrate

  • MDL number MFCD02260485
  • Cas Number : 890100-63-7
  • Molecular Weight : 241.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5536B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5536B
Availability details for
Quantity
  • Availability
  • Add to Cart