RIE156321026 Rieke Metals, Inc.

Ethyl 7-(6-chloropyridin-3-yl)-7-oxoheptanoate

  • MDL number MFCD02260488
  • Cas Number : 890100-69-3
  • Molecular Weight : 283.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5536E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5536E
Availability details for
Quantity
  • Availability
  • Add to Cart