RIE156321027 Rieke Metals, Inc.

Ethyl 8-(6-chloropyridin-3-yl)-8-oxooctanoate

  • MDL number MFCD02260489
  • Cas Number : 890100-72-8
  • Molecular Weight : 297.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5536F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5536F
Availability details for
Quantity
  • Availability
  • Add to Cart