RIE156321029 Rieke Metals, Inc.

5-(2-Chloro-3-pyridyl)-5-oxovaleronitrile

  • MDL number MFCD02260496
  • Cas Number : 890100-76-2
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5538C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5538C
Availability details for
Quantity
  • Availability
  • Add to Cart