RIE156321030 Rieke Metals, Inc.

6-(2-Chloro-3-pyridyl)-6-oxohexanenitrile

  • MDL number MFCD02260497
  • Cas Number : 890100-78-4
  • Molecular Weight : 222.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5538D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5538D
Availability details for
Quantity
  • Availability
  • Add to Cart