RIE156321031 Rieke Metals, Inc.

7-(2-Chloro-3-pyridyl)-7-oxoheptanenitrile

  • MDL number MFCD02260498
  • Cas Number : 890100-80-8
  • Molecular Weight : 236.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5538E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5538E
Availability details for
Quantity
  • Availability
  • Add to Cart