RIE156321032 Rieke Metals, Inc.

8-(2-Chloro-3-pyridyl)-8-oxooctanenitrile

  • MDL number MFCD02260499
  • Cas Number : 890100-82-0
  • Molecular Weight : 250.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5538F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5538F
Availability details for
Quantity
  • Availability
  • Add to Cart