RIE156321033 Rieke Metals, Inc.

4-(4-chloro-3-pyridyl)-4-oxobutyronitrile

  • MDL number MFCD07783633
  • Cas Number : 890100-84-2
  • Molecular Weight : 194.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5539B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5539B
Availability details for
Quantity
  • Availability
  • Add to Cart