RIE156321035 Rieke Metals, Inc.

6-(4-chloro-3-pyridyl)-6-oxohexanenitrile

  • MDL number MFCD07783635
  • Cas Number : 890100-88-6
  • Molecular Weight : 222.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5539D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5539D
Availability details for
Quantity
  • Availability
  • Add to Cart