RIE156321036 Rieke Metals, Inc.

7-(4-chloro-3-pyridyl)-7-oxoheptanenitrile

  • MDL number MFCD07783636
  • Cas Number : 890100-90-0
  • Molecular Weight : 236.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5539E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5539E
Availability details for
Quantity
  • Availability
  • Add to Cart