RIE156321037 Rieke Metals, Inc.

8-(4-chloro-3-pyridyl)-8-oxooctanenitrile

  • MDL number MFCD07783637
  • Cas Number : 890100-92-2
  • Molecular Weight : 250.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5539F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5539F
Availability details for
Quantity
  • Availability
  • Add to Cart