RIE156321038 Rieke Metals, Inc.

2-Chloro-3-(5-chlorovaleryl)pyridine

  • MDL number MFCD02260512
  • Cas Number : 914203-36-4
  • Molecular Weight : 232.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-1-(2-chloro-3-pyridinyl)-1-pentanone
Inchi Code 1S/C10H11Cl2NO/c11-6-2-1-5-9(14)8-4-3-7-13-10(8)12/h3-4,7H,1-2,5-6H2
Inchi Key AKDNHMWBTVCIHY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5541d

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5541d
Availability details for
Quantity
  • Availability
  • Add to Cart